Learn More
Biomatik Corporation Human Sialic Acid Binding Ig Like Lectin 9 (SIGLEC9) ELISA Kit
Supplier:  Biomatik Corporation EKU12518
Encompass_Preferred
Human Sialic Acid Binding Ig Like Lectin 9 (SIGLEC9) ELISA Kit for detecting SIGLEC9 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y336. Gene ID: 27180. Target Synonyms: CDw329; CD329; OBBP-LIKE; Protein FOAP-9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6694