Learn More
Biomatik Corporation Human Sialic Acid Binding Ig Like Lectin 7 (SIGLEC7) ELISA Kit
Supplier:  Biomatik Corporation EKU09084
Encompass_Preferred
Human Sialic Acid Binding Ig Like Lectin 7 (SIGLEC7) ELISA Kit for detecting SIGLEC7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y286. Gene ID: 27036. Target Synonyms: CDw328; CD328; p75; AIRM1; D-Siglec; QA79; SIGLEC-7; P75/AIRM1; Adhesion inhibitory receptor molecule 1; QA79 membrane protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4447