Learn More
Biomatik Corporation Human Sialyltransferase 1 (SIAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU11827
Encompass_Preferred
Human Sialyltransferase 1 (SIAT1) ELISA Kit for detecting SIAT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15907. Gene ID: 6480. Target Synonyms: CD75; ST6GAL1; ST6Gal I; ST6GalI; B-cell antigen CD75; Beta-galactoside alpha-2,6-sialyltransferase 1; ST6 Beta Galactosamide Alpha-2,6-Sialyltranferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7058