Learn More
Sino Biological Anti-SIGIRR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12165RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SIGIRR (rh SIGIRR; Catalog#12165-H08H; Met 1-His 118; Q6IA17-1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4437