Learn More
Sino Biological Anti-SIGIRR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12165RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SIGIRR (rh SIGIRR; Catalog#12165-H08H; Met 1-His 118; Q6IA17-1). SIGIRR specific IgG was purified by Human SIGIRR affinity chromatography.
Catalog No. 50-255-4436