Learn More
Sino Biological Recombinant Human SIGIRR Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 12165H08H1
Encompass_Preferred
A DNA sequence encoding the human SIGIRR (Q6IA17-1) extracellular domain (Met 1-His 118) was expressed, fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-224-6900