Learn More
ACROBiosystems PE-Labeled Human Siglec-10 Protein, His Tag (recommended for neutralizing assay)
Catalog No. 502331648
Encompass_Preferred
PE-Labeled Human Siglec-10, His Tag (SI0-HP2E5) is produced via site-specific conjugation of PE to Human Siglec-10, His Tag under optimal conditions with a proprietary technology with a proprietary technology. Human Siglec-10, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).
Catalog No. 50-233-1648 Supplier ACROBiosystems Supplier No. SI0HP2E550TESTS
PE-Labeled Human Siglec-10, His Tag (SI0-HP2E5) is produced via site-specific conjugation of PE to Human Siglec-10, His Tag under optimal conditions with a proprietary technology with a proprietary technology. Human Siglec-10, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).