Learn More
Bon Opus Biosciences Recombinant Human SIGLEC-10 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP105650UG
Encompass_Preferred
Recombinant Human SIGLEC-10 (C-hFc) is produced by Human Cells. The target gene encoding Met 17-Thr 546 is expressed with a C-hFc tag. Accession# Q96LC7-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1056
Catalog No. 50-217-2654