Learn More
ACROBiosystems ACROBiosystems Human Siglec-10 Protein, Mouse IgG2a Fc Tag, low endotoxin
Catalog No. 502106670
Encompass_Preferred
Recombinant Protein;1MG;Human Siglec-10, Mouse IgG2a Fc Tag, low endotoxin (SI0-H525b) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 85.6 kDa. The protein migrates as 90-120 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-10, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SI0-H525b) at 5 μg/mL (100 μL/well) can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0.6-20 ng/mL (QC tested).
Catalog No. 50-210-6670 Supplier ACROBiosystems Supplier No. SI0H525B1MG
Recombinant Protein;1MG;Human Siglec-10, Mouse IgG2a Fc Tag, low endotoxin (SI0-H525b) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 85.6 kDa. The protein migrates as 90-120 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-10, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SI0-H525b) at 5 μg/mL (100 μL/well) can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0.6-20 ng/mL (QC tested).