Learn More
ACROBiosystems Human Siglec-2 200tests
Catalog No. 502534178
Encompass_Preferred
APC-Labeled Human Siglec-2 Protein His Tag (SI2-HA2H4) is produced via conjugation of APC to Human Siglec-2 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-2 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).
Catalog No. 50-253-4178 Supplier ACROBiosystems Supplier No. SI2HA2H4200TESTS
APC-Labeled Human Siglec-2 Protein His Tag (SI2-HA2H4) is produced via conjugation of APC to Human Siglec-2 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-2 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).