Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human Siglec-2 Protein, Fc Tag, Low Endotoxin, 1 mg

Catalog No. 502107292
Encompass_Preferred

Human Siglec-2 (CD22) is a member of the immunoglobulin superfamily and the SIGLEC family. This protein is expressed in HEK293 cells and carries a human IgG1 Fc tag at the C-terminus, containing amino acids Asp 20 to Arg 687. It functions as an inhibitory receptor, mediating B-cell interactions and signal transduction through the recruitment of cytoplasmic phosphatases. This low endotoxin protein is verified by MALS.

  • Purity: ≥95% by SDS-PAGE and SEC-MALS.
  • Calculated MW: 101.6 kDa, migrating as 125-150 kDa due to glycosylation.
  • Endotoxin: <0.01 EU per μg by LAL/rFC method.
  • Formulation: Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH 7.5 with trehalose.
  • Recommended storage: -20°C to -70°C for 12 months in lyophilized state.

Catalog No. 50-210-7292 Supplier ACROBiosystems Supplier No. CD2H52531MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Siglec-2 (CD22) is a member of the immunoglobulin superfamily and the SIGLEC family. This protein is expressed in HEK293 cells and carries a human IgG1 Fc tag at the C-terminus, containing amino acids Asp 20 to Arg 687. It functions as an inhibitory receptor, mediating B-cell interactions and signal transduction through the recruitment of cytoplasmic phosphatases. This low endotoxin protein is verified by MALS.

  • Purity: ≥95% by SDS-PAGE and SEC-MALS.
  • Calculated MW: 101.6 kDa, migrating as 125-150 kDa due to glycosylation.
  • Endotoxin: <0.01 EU per μg by LAL/rFC method.
  • Formulation: Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH 7.5 with trehalose.
  • Recommended storage: -20°C to -70°C for 12 months in lyophilized state.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.