Learn More
ACROBiosystems APC-Labeled Human Siglec-3 / CD33 Protein, His TagStar Staining
Catalog No. 502332041
Encompass_Preferred
APC-Labeled Human Siglec-3 Protein, His Tag (CD3-HA2H3) is produced via conjugation of APC to Human Siglec-3 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).
Catalog No. 50-233-2041 Supplier ACROBiosystems Supplier No. CD3HA2H325TESTS
APC-Labeled Human Siglec-3 Protein, His Tag (CD3-HA2H3) is produced via conjugation of APC to Human Siglec-3 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).