Learn More
ACROBiosystems PE-Labeled Human Siglec-3 / CD33 Protein, His TagStar Staining
Catalog No. 502331513
Encompass_Preferred
PE-Labeled Human Siglec-3 Protein, His Tag (CD3-HP2H7) is produced via conjugation of PE to Human Siglec-3 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).
Catalog No. 50-233-1513 Supplier ACROBiosystems Supplier No. CD3HP2H725TESTS
PE-Labeled Human Siglec-3 Protein, His Tag (CD3-HP2H7) is produced via conjugation of PE to Human Siglec-3 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-3 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).