Learn More
ACROBiosystems Alexa Fluor 647-Labeled Human Siglec-3 / CD33 Protein, His TagStar Staining
Catalog No. 502331583
Encompass_Preferred
Alexa Fluor 647-Labeled Human Siglec-3, His Tag (CD3-HA2H6) is produced via conjugation of AF647 to Human Siglec-3, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-3, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).
Catalog No. 50-233-1583 Supplier ACROBiosystems Supplier No. CD3HA2H625UG
Alexa Fluor 647-Labeled Human Siglec-3, His Tag (CD3-HA2H6) is produced via conjugation of AF647 to Human Siglec-3, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Siglec-3, His Tag is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).