Learn More
ACROBiosystems ACROBiosystems Human Siglec-3 / CD33 Protein, Llama IgG2b Fc Tag, low endotoxin
Catalog No. 502106548
Encompass_Preferred
Recombinant Protein;100UG;Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin (CD3-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 54.6 kDa. The protein migrates as 64-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. CD3-H5259) at 5 μg/mL (100 μL/well) can bind Anti-CD33, Human IgG1 with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-210-6548 Supplier ACROBiosystems Supplier No. CD3H5259100UG
Recombinant Protein;100UG;Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin (CD3-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 54.6 kDa. The protein migrates as 64-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. CD3-H5259) at 5 μg/mL (100 μL/well) can bind Anti-CD33, Human IgG1 with a linear range of 0.078-1.25 μg/mL (QC tested).