Learn More
Raybiotech Inc Human Siglec-3 ELISA Kit
Supplier:  Raybiotech Inc ELHSIGLEC31
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Human Siglec-3. Other species not determined.
Catalog No. 50-194-9727