Learn More
Sino Biological Human Siglec-5 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786612
Encompass_Preferred
The Human SIGLEC5 ELISA Pair Set is for the quantitative determination of Human SIGLEC5.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human SIGLEC5 coated on a 96-well plate. Standards and samples are added to the wells, and any Human SIGLEC5 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human SIGLEC5 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human SIGLEC5 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86612 Supplier Sino Biological Supplier No. SEK1179815
The Human SIGLEC5 ELISA Pair Set is for the quantitative determination of Human SIGLEC5.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human SIGLEC5 coated on a 96-well plate. Standards and samples are added to the wells, and any Human SIGLEC5 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human SIGLEC5 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human SIGLEC5 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.