Learn More
Bon Opus Biosciences Recombinant Human SIGLEC-6 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP105050UG
Encompass_Preferred
Recombinant Human SIGLEC-6 (C-hFc) is produced by Human Cells. The target gene encoding Gln27-Val331 is expressed with a C-hFc tag. Accession# AAH35359. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1050
Catalog No. 50-217-2648