Learn More
Bon Opus Biosciences Recombinant Human SIGLEC-6 (C-mFc) 50ug
Supplier: Bon Opus Biosciences BP105150UG
Recombinant Human SIGLEC-6 (C-mFc) is produced by Human Cells. The target gene encoding Gln27-Val331 is expressed with a C-mFc tag. Accession# AAH35359. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1051