Learn More
ACROBiosystems Human Siglec-6 Protein, Fc Tag, Low Endotoxin, 100 μg
Catalog No. 502106768
Encompass_Preferred
Human Siglec-6, also known as CD327, is a member of the immunoglobulin superfamily and the sialic acid-binding Ig-like lectin family. It features two Ig-like C2-type domains and one Ig-like V-type domain, mediating sialic acid-dependent binding to cells. This protein is expressed in human HEK293 cells and contains the extracellular domain (Gln 16 to Val 320) fused to a human IgG1 Fc tag at the C-terminus.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 60.3 kDa, migrating at 80–100 kDa under reducing conditions
  • Low endotoxin levels, less than 0.01 EU per μg
  • Lyophilized formulation in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7.5, with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state
Catalog No. 50-210-6768 Supplier ACROBiosystems Supplier No. SI6H5256100UG
Human Siglec-6, also known as CD327, is a member of the immunoglobulin superfamily and the sialic acid-binding Ig-like lectin family. It features two Ig-like C2-type domains and one Ig-like V-type domain, mediating sialic acid-dependent binding to cells. This protein is expressed in human HEK293 cells and contains the extracellular domain (Gln 16 to Val 320) fused to a human IgG1 Fc tag at the C-terminus.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 60.3 kDa, migrating at 80–100 kDa under reducing conditions
  • Low endotoxin levels, less than 0.01 EU per μg
  • Lyophilized formulation in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7.5, with trehalose
  • Stable for 12 months at -20°C to -70°C in lyophilized state