Learn More
ACROBiosystems Biotinylated Human Siglec-7, His,Avitag (SG7-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Leu 353 (Accession # Q9Y286-1).
Supplier:  ACROBiosystems SG7H82E725UG
Encompass_Preferred
Siglec-7 is a member of the human CD33‐related Siglec receptor. The extracellular region of Siglec‐7 is characterized by an N‐terminal V‐set Ig domain that can bind sialic acid and two C2‐set Ig domains. The cytoplasmic tail of Siglec‐7 has one immune‐receptor tyrosine‐based inhibitory motif (ITIM) and one ITIM‐like motif. Siglec-7 is considered as a sialic acid-dependent immunoreceptor with inhibitory potential and expressed predominantly on human NK cells, monocytes and a small subset of CD8+ T cells.
Catalog No. 50-223-0847