Learn More
ACROBiosystems Human Siglec-7, Fc (L118A, G120A, E201A) Tag (SI7-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Leu 353 (Accession # Q9Y286-1).
Catalog No. 502253161
Encompass_Preferred
Siglec-7 is a member of the human CD33‐related Siglec receptor. The extracellular region of Siglec‐7 is characterized by an N‐terminal V‐set Ig domain that can bind sialic acid and two C2‐set Ig domains. The cytoplasmic tail of Siglec‐7 has one immune‐receptor tyrosine‐based inhibitory motif (ITIM) and one ITIM‐like motif. Siglec-7 is considered as a sialic acid-dependent immunoreceptor with inhibitory potential and expressed predominantly on human NK cells, monocytes and a small subset of CD8+ T cells.
This item is not returnable. View return policy
Catalog No. 50-225-3161 Supplier ACROBiosystems Supplier No. SI7H525750UG
Siglec-7 is a member of the human CD33‐related Siglec receptor. The extracellular region of Siglec‐7 is characterized by an N‐terminal V‐set Ig domain that can bind sialic acid and two C2‐set Ig domains. The cytoplasmic tail of Siglec‐7 has one immune‐receptor tyrosine‐based inhibitory motif (ITIM) and one ITIM‐like motif. Siglec-7 is considered as a sialic acid-dependent immunoreceptor with inhibitory potential and expressed predominantly on human NK cells, monocytes and a small subset of CD8+ T cells.