Learn More
Sino Biological Recombinant Human Siglec-10 Protein (ECD, mFc Tag), HPLC-verified
Supplier:  Sino Biological 29666H38H50
Encompass_Preferred
A DNA sequence encoding the human SIGLEC10 (NP_149121.2) (Met17-Thr546) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-3825