Learn More
Sino Biological Recombinant Human Siglec-15 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 13976H02H150
Encompass_Preferred
A DNA sequence encoding the human SIGLEC15 (NP_998767.1) (Met1-Thr263) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4150