Learn More
Sino Biological Human SIGLEC5 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11798MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SIGLEC5 (rh SIGLEC5; Catalog#11798-HCCH; O15389; Met1-Thr 434). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1010