Learn More
BIOHIPPO Human Siglec5 Microsample
SDP
Catalog No. 5000313999
Encompass
Human Siglec5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Siglec5 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human Siglec5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human Siglec5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human Siglec5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13999 Supplier BIOHIPPO Supplier No. BHE15212386
Human Siglec5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Siglec5 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human Siglec5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human Siglec5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human Siglec5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color