Learn More
Sino Biological Recombinant Human Siglec-7 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 30108H08H500
Encompass_Preferred
A DNA sequence encoding the Human SIGLEC7 (Q9Y286-1) (Met1-Leu353) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-1238