Learn More
Biomatik Corporation Human Signal Recognition Particle Receptor (SRPR) ELISA Kit
Supplier:  Biomatik Corporation EKU11772
Encompass_Preferred
Human Signal Recognition Particle Receptor (SRPR) ELISA Kit for detecting SRPR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08240. Gene ID: 6734. Target Synonyms: DP; SRP-alpha; Sralpha; Docking Protein; Signal recognition particle receptor subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7630