Learn More
Biomatik Corporation Human Signal Regulatory Protein Gamma (SIRPg) ELISA Kit
Supplier:  Biomatik Corporation EKU07334
Encompass_Preferred
Human Signal Regulatory Protein Gamma (SIRPg) ELISA Kit for detecting SIRPg in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P1W8. Gene ID: 55423. Target Synonyms: CD172g; CD172-G; SIRP-G; SIRP-B2; SIRPB2; SIRP-b2; SIRP-beta-2; SIRPgamma; CD172 antigen-like family member B; Signal-regulatory protein beta-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7361