Learn More
Biomatik Corporation Human Signaling Lymphocytic Activation Molecule Family, Member 2 (SLAMF2) ELISA Kit
Supplier:  Biomatik Corporation EKU11977
Encompass_Preferred
Human Signaling Lymphocytic Activation Molecule Family, Member 2 (SLAMF2) ELISA Kit for detecting SLAMF2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09326. Gene ID: 962. Target Synonyms: CD48; BLAST; BLAST1; MEM-102; BCM1; HCD48; MCD48; B-lymphocyte activation marker BLAST-1; BCM1 surface antigen; Leukocyte antigen MEM-102; SLAM family member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6242