Learn More
BIOHIPPO Human sIL-6R Microsample
SDP
Catalog No. 5000315902
Encompass
Human sIL-6R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sIL-6R in Serum plasma tissue homogenates urine and other biological fluids It provides a detection range of 1 88-120 ng/mL with a sensitivity of 0 78 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sIL-6R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sIL-6R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sIL-6R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15902 Supplier BIOHIPPO Supplier No. BHE15212078
Human sIL-6R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sIL-6R in Serum plasma tissue homogenates urine and other biological fluids It provides a detection range of 1 88-120 ng/mL with a sensitivity of 0 78 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sIL-6R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sIL-6R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sIL-6R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color