Learn More
BIOHIPPO Human sIL2Ra Microsample
SDP
Catalog No. 5000313996
Encompass
Human sIL2Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sIL2Ra in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 18 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sIL2Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sIL2Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sIL2Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13996 Supplier BIOHIPPO Supplier No. BHE15212368
Human sIL2Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sIL2Ra in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 18 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sIL2Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sIL2Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sIL2Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color