Learn More
Biomatik Corporation Human Single Strand Selective Monofunctional Uracil DNA Glycosylase 1 (SMUG1) ELISA Kit
Supplier:  Biomatik Corporation EKU07347
Encompass_Preferred
Human Single Strand Selective Monofunctional Uracil DNA Glycosylase 1 (SMUG1) ELISA Kit for detecting SMUG1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.051ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q53HV7. Gene ID: 23583. Target Synonyms: FDG; HMUDG; UNG3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8757