Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SIPA1L2 Synthetic Peptide

Catalog No. PEP0962 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0962 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0962 Supplier Invitrogen™ Supplier No. PEP0962
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 19 amino acids near the amino terminus of human SIPA1L2. PEP-0962 can be used as a blocking peptide with polyclonal antibody PA5-20848.

Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 57568
Species Human
Purification Method purified
Accession Number Q9P2F8
Common Name SIPA1L2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias BC058408; Kiaa0545; KIAA1389; mKIAA1389; serine-rich synapse associated protein 2; serine-rich synapse-associated protein; Sersap2; signal induced proliferation associated 1 like 2; signal-induced proliferation-associated 1 like 2; signal-induced proliferation-associated 1-like protein 2; Sipa1l2; SIPA1-like protein 2; SPA-1-like 2; SPAL2; Spar2
Gene Symbol SIPA1L2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.