Learn More
ACROBiosystems ACROBiosystems Human SIRP alpha / CD172a Protein, Fc Tag (MALS verified)
Catalog No. 502106767
Encompass_Preferred
Recombinant Protein;1MG;Human SIRP alpha, Fc Tag (SIA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession # NP_001035111).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.9 kDa. The protein migrates as 70-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 2 μg/mL (100 μL/well) can bind Human CD47, His Tag (Cat. No. CD7-H5227) with a linear range of 2-13 ng/mL (QC tested).;FACS assay shows that Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) can bind to Jurkat cell expressing CD47. The concentration of SIRP alpha used is 1 μg/mL (Routinely tested).
Catalog No. 50-210-6767 Supplier ACROBiosystems Supplier No. SIAH52511MG
Recombinant Protein;1MG;Human SIRP alpha, Fc Tag (SIA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession # NP_001035111).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.9 kDa. The protein migrates as 70-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 2 μg/mL (100 μL/well) can bind Human CD47, His Tag (Cat. No. CD7-H5227) with a linear range of 2-13 ng/mL (QC tested).;FACS assay shows that Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) can bind to Jurkat cell expressing CD47. The concentration of SIRP alpha used is 1 μg/mL (Routinely tested).