Learn More
ACROBiosystems Human SIRP alphaV2 / CD172a Protein, His Tag
Catalog No. 502055047
Encompass_Preferred
Recombinant Protein;1MG;Human SIRP alphaV2 Protein, His Tag (SI2-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 369 (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (HPLC-verified) (Cat. No. CD7-H5256) at 2 μg/mL (100 μL/well) can bind Human SIRP alphaV2 Protein, His Tag (Cat. No. SI2-H52H9) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-205-5047 Supplier ACROBiosystems Supplier No. SI2H52H91MG
Recombinant Protein;1MG;Human SIRP alphaV2 Protein, His Tag (SI2-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 369 (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (HPLC-verified) (Cat. No. CD7-H5256) at 2 μg/mL (100 μL/well) can bind Human SIRP alphaV2 Protein, His Tag (Cat. No. SI2-H52H9) with a linear range of 0.078-1.25 μg/mL (QC tested).