Learn More
ACROBiosystems ACROBiosystems Human SIRP alphaV8 / CD172a Protein, His Tag (MALS verified)
Catalog No. 502106457
Encompass_Preferred
Recombinant Protein;1MG;Human SIRP alphaV8, His Tag (SI8-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 369 (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.1 kDa. The protein migrates as 46-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alphaV8, His Tag (Cat. No. SI8-H52H5) at 2 μg/mL (100 μL/well) can bind Human CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H52A5) with a linear range of 1-5 ng/mL (QC tested).
Catalog No. 50-210-6457 Supplier ACROBiosystems Supplier No. SI8H52H51MG
Recombinant Protein;1MG;Human SIRP alphaV8, His Tag (SI8-H52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 369 (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.1 kDa. The protein migrates as 46-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alphaV8, His Tag (Cat. No. SI8-H52H5) at 2 μg/mL (100 μL/well) can bind Human CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H52A5) with a linear range of 1-5 ng/mL (QC tested).