Learn More
Sino Biological Recombinant Human SIRP alpha/SHPS-1 Protein (ECD, SIRP alphaV8, mFc Tag)
Supplier:  Sino Biological 30015H38H100
Encompass_Preferred
A DNA sequence encoding the human SIRP alphaV8 (AAH38510.1, with mutation Ser 44 Leu, Ser 50 Thr, Ile 52 Thr, His 54 Arg, Val 57 Ala) (Met1-Arg369) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-3809