Learn More
Sino Biological Recombinant Human SIRP alpha/SHPS-1 Protein (ECD, SIRP alphaV2, mFc Tag)
Supplier:  Sino Biological 30014H38H100
Encompass_Preferred
A DNA sequence encoding the human SIRP alphaV2 (AAH38510.1) (Met1-Arg369) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-3811