Learn More
Sino Biological Recombinant Human SIRP alpha/SHPS-1 Protein (G75A, ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11612H08H1
Encompass_Preferred
A DNA sequence encoding the human SIRPA (NP_542970.1, with natural variant Gly 75 Ala) extracellular domain (Met1-Arg370) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4665