Learn More
Bon Opus Biosciences Recombinant Human SIRP alpha/CD172a (C-His) - Source: Human 293 Cells
SDP
Supplier:  Bon Opus Biosciences BP22950UG
Encompass_Preferred
Recombinant Human SIRP alpha is produced by our Mammalian expression system and the target gene encoding Glu31-Arg370 is expressed with a 6His tag at the C-terminus. Accession#P78324. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4
Catalog No. NC2733668