Learn More
Sino Biological Anti-SIRP gamma/SIRPG Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11828MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SIRPG / SIRP gamma / CD172g (rh SIRPG / SIRP gamma / CD172g; Catalog#11828-H08H; NP_061026.2; Met 1-Ser 364). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4531