Learn More
Sino Biological Recombinant Human SIRP gamma/SIRPG Protein (hFc Tag)
Supplier:  Sino Biological 11828H02H1
Encompass_Preferred
A DNA sequence encoding the human SIRPG ( NP_061026.2) (Met1-Ser364) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-241-1431