Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SKIIP Partial ORF (NP_036377, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7099327 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-999 25 μg
89-961-998 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-999 Supplier Abnova™ Supplier No. H00022938Q01L

Used for AP, Array, ELISA, WB-Re

This gene, a member of the SNW gene family, encodes a coactivator that enhances transcription from some Pol II promoters. This coactivator can bind to the ligand-binding domain of the vitamin D receptor and to retinoid receptors to enhance vitamin D-, retinoic acid-, estrogen-, and glucocorticoid-mediated gene expression. It can also function as a splicing factor by interacting with poly(A)-binding protein 2 to directly control the expression of muscle-specific genes at the transcriptional level. Finally, the protein may be involved in oncogenesis since it interacts with a region of SKI oncoproteins that is required for transforming activity. [provided by RefSeq]

Sequence: MALTSFLPAPTQLSQDQLEAEEKARSQRSRQTSLVSSRREPPPYGYRKGWIPRLLEDFGDGGAFPEIHVAQYPLDMGRKKKMSNALAIQVDSEGKIKYDAIARQGQSKDK

Specifications

Accession Number NP_036377
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 22938
Molecular Weight (g/mol) 37.84kDa
Name SKIIP (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MALTSFLPAPTQLSQDQLEAEEKARSQRSRQTSLVSSRREPPPYGYRKGWIPRLLEDFGDGGAFPEIHVAQYPLDMGRKKKMSNALAIQVDSEGKIKYDAIARQGQSKDK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Bx42/MGC119379/NCOA-62/PRPF45/Prp45/SKIIP/SKIP
Common Name SNW1
Gene Symbol SNW1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.