Learn More
Biomatik Corporation Human Skin Aspartic Protease (SASP) ELISA Kit
Supplier:  Biomatik Corporation EKU12095
Encompass_Preferred
Human Skin Aspartic Protease (SASP) ELISA Kit for detecting SASP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q53RT3. Gene ID: 151516. Target Synonyms: ASPRV1; Taps; SASPase; MUNO; Aspartic Peptidase,Retroviral-Like 1; Skin-specific retroviral-like aspartic protease; TPA-inducible aspartic proteinase-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6783