Learn More
Sino Biological Anti-SLAM/CD150 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10837RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SLAMF1 / CD150 (rhSLAMF1; Catalog#10837-H08H; NP_003028.1; Met 1-Pro 258). SLAMF1 specific IgG was purified by human SLAMF1 affinity chromatography.
Catalog No. 50-240-8734