Learn More
Sino Biological Human SLAMF1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10837T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD150/SLAM/SLAMF1 (rh CD150/SLAM/SLAMF1; Catalog#10837-H08H; NP_003028.1; Met1-Pro258). CD150/SLAM/SLAMF1 specific IgG was purified by Human CD150/SLAM/SLAMF1 affinity chromatography.
Catalog No. 50-237-3996