Learn More
ACROBiosystems ACROBiosystems Human SLAMF1 / SLAM / CD150 Protein, His Tag
Supplier:  ACROBiosystems SL1H52H0100UG
Encompass_Preferred
Recombinant Protein;100UG;Human SLAMF1, His Tag (SL1-H52H0) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Pro 237 (Accession # Q13291-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6725