Learn More
Sino Biological Recombinant Human SLAMF6 Protein (hFc Tag) 1
Supplier:  Sino Biological 11945H02H1
Encompass_Preferred
A DNA sequence encoding the human SLAMF6 (Q96DU3-1) (Met1-Met226) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7089