Learn More
Sino Biological Recombinant Anti-SLAMF6 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 11945R002F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Ly108/SLAMF6 (rh Ly108/SLAMF6; Catalog#11945-H08H; Q96DU3-1; Met1-Met226) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8510